siRNA Details
Gene Symbol: LOC100912585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246555.2 | XP_006246617.1 | 4 | 869 | ||
XM_006246556.2 | XP_006246618.1 | 4 | 1332 | |||
XM_006246558.2 | XP_006246620.1 | 3 | 405 | |||
XM_006246559.2 | XP_006246621.1 | 3 | 377 | |||
XM_008767892.1 | XP_008766114.1 | 3 | 994 | |||
XM_008767893.1 | XP_008766115.1 | 3 | 427 |
Gene Symbol: Mapk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191547.1 | NP_001178476.1 | 550 | |||
XM_006246417.2 | XP_006246479.1 | 869 | ||||
XM_006246418.2 | XP_006246480.1 | 1332 | ||||
XM_006246420.2 | XP_006246482.1 | 405 | ||||
XM_006246421.2 | XP_006246483.1 | 377 | ||||
XM_008767756.1 | XP_008765978.1 | 427 |
