siRNA Details
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053720.1 | NP_446172.1 | 12 | 1597 | ||
GenBank mRNA | AJ238717.3 | - | 12 | 1597 | ||
BC078769.1 | - | 12 | 1574 | 7110171 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053720.1 | NP_446172.1 | 12 | 1597 | ||
GenBank mRNA | AJ238717.3 | - | 12 | 1597 | ||
BC078769.1 | - | 12 | 1574 | 7110171 |