siRNA Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053720.1 | NP_446172.1 | 1 | 172 | ||
| GenBank mRNA | AJ238717.3 | - | 1 | 172 | ||
| BC078769.1 | - | 1 | 149 | 7110171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aatf| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053720.1 | NP_446172.1 | 1 | 172 | ||
| GenBank mRNA | AJ238717.3 | - | 1 | 172 | ||
| BC078769.1 | - | 1 | 149 | 7110171 | ||