siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053723.1 | NP_446175.1 | 3, 4 | 555 | ||
| GenBank mRNA | AJ131111.1 | - | 3, 4 | 555 | ||
| AJ295233.1 | - | 3, 4 | 563 | |||
| BC085811.1 | - | 3, 4 | 533 | 7115423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lancl1| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053723.1 | NP_446175.1 | 3, 4 | 555 | ||
| GenBank mRNA | AJ131111.1 | - | 3, 4 | 555 | ||
| AJ295233.1 | - | 3, 4 | 563 | |||
| BC085811.1 | - | 3, 4 | 533 | 7115423 | ||