siRNA Details
| Gene Symbol: Tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053729.1 | NP_446181.1 | 9 | 1108 | ||
| GenBank mRNA | AY026945.1 | - | 9 | 1108 | ||
| BC129088.1 | - | 9 | 1125 | 8362408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdg| Gene Symbol: Tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053729.1 | NP_446181.1 | 9 | 1108 | ||
| GenBank mRNA | AY026945.1 | - | 9 | 1108 | ||
| BC129088.1 | - | 9 | 1125 | 8362408 | ||