siRNA Details
| Gene Symbol: Ncf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053734.2 | NP_446186.2 | 8 | 772 | ||
| GenBank mRNA | AF260779.1 | - | 7 | 700 | ||
| AF547392.1 | - | 8 | 772 | |||
| AF547393.1 | - | 8 | 772 | |||
| AY029167.1 | - | 8 | 772 | |||
| BC061810.1 | - | 8 | 772 | 5598653 | ||
| Gene Symbol: LOC100912379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769176.1 | XP_008767398.1 | 1 | 225 | ||

Ncf1
Genomic Map