siRNA Details
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053734.2 | NP_446186.2 | 8 | 722 | ||
GenBank mRNA | AF260779.1 | - | 7 | 650 | ||
AF547392.1 | - | 8 | 722 | |||
AF547393.1 | - | 8 | 722 | |||
AY029167.1 | - | 8 | 722 | |||
BC061810.1 | - | 8 | 722 | 5598653 |
Gene Symbol: LOC100912379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769176.1 | XP_008767398.1 | 1 | 175 |
