siRNA Details
| Gene Symbol: Wif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053738.1 | NP_446190.1 | 8 | 1162 | ||
| XM_006241395.2 | XP_006241457.1 | 8 | 1177 | |||
| GenBank mRNA | AY030278.1 | - | 8 | 1162 | ||
| BC072473.1 | - | 8 | 988 | 7100360 | ||

Wif1
Genomic Map