siRNA Details
| Gene Symbol: Cdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053743.1 | NP_446195.1 | 3 | 532 | ||
| GenBank mRNA | AB097113.1 | - | 3 | 445 | ||
| BC061720.1 | - | 3 | 501 | 5598479 | ||
| D26564.1 | - | 3 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc37| Gene Symbol: Cdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053743.1 | NP_446195.1 | 3 | 532 | ||
| GenBank mRNA | AB097113.1 | - | 3 | 445 | ||
| BC061720.1 | - | 3 | 501 | 5598479 | ||
| D26564.1 | - | 3 | 532 | |||