siRNA Details
| Gene Symbol: Dlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053744.1 | NP_446196.1 | 458 | |||
| XM_008764917.1 | XP_008763139.1 | 514 | ||||
| GenBank mRNA | BC167752.1 | - | 478 | 8365650 | ||
| D84336.1 | - | 458 | ||||
| U25680.1 | - | 323 | ||||

Dlk1
Genomic Map