siRNA Details
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053746.1 | NP_446198.1 | 17 | 2376 | ||
XM_008765902.1 | XP_008764124.1 | 17 | 2441 | |||
GenBank mRNA | D87212.1 | - | 17 | 2376 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053746.1 | NP_446198.1 | 17 | 2376 | ||
XM_008765902.1 | XP_008764124.1 | 17 | 2441 | |||
GenBank mRNA | D87212.1 | - | 17 | 2376 |