siRNA Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053747.2 | NP_446199.2 | 5 | 919 | ||
| XM_006253551.2 | XP_006253613.1 | 5 | 967 | |||
| GenBank mRNA | BC072477.1 | - | 5 | 919 | 7103662 | |
| FQ222353.1 | - | 5 | 946 | |||

Ubqln1
Genomic Map