siRNA Details
| Gene Symbol: Ddx39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133300.3 | NP_579834.2 | 5 | 743 | ||
| GenBank mRNA | BC080243.1 | - | 5 | 692 | 7100155 | |
| FQ212901.1 | - | 5 | 729 | |||
| M75168.1 | - | 4 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx39b| Gene Symbol: Ddx39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133300.3 | NP_579834.2 | 5 | 743 | ||
| GenBank mRNA | BC080243.1 | - | 5 | 692 | 7100155 | |
| FQ212901.1 | - | 5 | 729 | |||
| M75168.1 | - | 4 | 648 | |||