siRNA Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053755.2 | NP_446207.1 | 13 | 1642 | ||
| GenBank mRNA | AF314820.1 | - | 10 | 1166 | ||
| AF337809.1 | - | 13 | 1647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a3| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053755.2 | NP_446207.1 | 13 | 1642 | ||
| GenBank mRNA | AF314820.1 | - | 10 | 1166 | ||
| AF337809.1 | - | 13 | 1647 | |||