siRNA Details
| Gene Symbol: Aimp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053757.1 | NP_446209.1 | 3 | 271 | ||
| GenBank mRNA | BC098675.1 | - | 3 | 271 | 7322792 | |
| FQ226969.1 | - | 1 | 47 | |||
| FQ229959.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aimp1| Gene Symbol: Aimp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053757.1 | NP_446209.1 | 3 | 271 | ||
| GenBank mRNA | BC098675.1 | - | 3 | 271 | 7322792 | |
| FQ226969.1 | - | 1 | 47 | |||
| FQ229959.1 | - | 1 | 47 | |||