siRNA Details
| Gene Symbol: Plce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053758.2 | NP_446210.1 | 24 | 5928 | ||
| XM_008760360.1 | XP_008758582.1 | 24 | 5892 | |||
| XM_008760361.1 | XP_008758583.1 | 24 | 7507 | |||
| GenBank mRNA | AF323615.1 | - | 24 | 5931 | ||

Plce1
Genomic Map