siRNA Details
| Gene Symbol: Gne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053765.2 | NP_446217.1 | 7 | 1436 | ||
| XM_006238006.2 | XP_006238068.1 | 7 | 1254 | |||
| XM_006238008.1 | XP_006238070.1 | 7 | 1220 | |||
| XM_006238009.2 | XP_006238071.1 | 7 | 1597 | |||
| GenBank mRNA | BC062011.1 | - | 7 | 1436 | 5621384 | |
| FQ224587.1 | - | 6 | 1099 | |||
| Y07744.1 | - | 6 | 1144 | |||

Gne
Genomic Map