siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053766.1 | NP_446218.1 | 5, 6 | 567 | ||
| XM_008770840.1 | XP_008769062.1 | 2, 3 | 1083 | |||
| XM_008770841.1 | XP_008769063.1 | 3, 4 | 586 | |||
| GenBank mRNA | BC089052.1 | - | 5, 6 | 567 | 7307299 | |
| FQ226684.1 | - | 4, 5 | 349 | |||

Wbp4
Genomic Map