siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053766.1 | NP_446218.1 | 4, 5 | 428 | ||
| GenBank mRNA | BC089052.1 | - | 4, 5 | 428 | 7307299 | |
| FQ226684.1 | - | 3, 4 | 210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbp4| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053766.1 | NP_446218.1 | 4, 5 | 428 | ||
| GenBank mRNA | BC089052.1 | - | 4, 5 | 428 | 7307299 | |
| FQ226684.1 | - | 3, 4 | 210 | |||