siRNA Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053767.1 | NP_446219.1 | 17 | 1815 | ||
| XM_006230449.2 | XP_006230511.1 | 19 | 4736 | |||
| XM_006230450.2 | XP_006230512.1 | 16 | 1743 | |||
| XM_006230451.2 | XP_006230513.1 | 17 | 4047 | |||
| XM_008759971.1 | XP_008758193.1 | 18 | 2414 | |||
| XM_008759972.1 | XP_008758194.1 | 18 | 2915 | |||
| GenBank mRNA | BC166573.1 | - | 17 | 1825 | 7320594 | |
| D78610.1 | - | 16 | 1689 | |||
| Y07834.1 | - | 2 | 209 | |||

Ptpre
Genomic Map