siRNA Details
| Gene Symbol: Wnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191848.1 | NP_001178777.1 | 3 | 420 | ||
| GenBank mRNA | AF204873.1 | - | 2 | 180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt2b| Gene Symbol: Wnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191848.1 | NP_001178777.1 | 3 | 420 | ||
| GenBank mRNA | AF204873.1 | - | 2 | 180 | ||