siRNA Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053786.1 | NP_446238.1 | 4 | 325 | ||
| GenBank mRNA | BC082110.1 | - | 4 | 325 | 7135071 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfc2| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053786.1 | NP_446238.1 | 4 | 325 | ||
| GenBank mRNA | BC082110.1 | - | 4 | 325 | 7135071 | |