siRNA Details
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053792.1 | NP_446244.1 | 25 | 2777 | ||
GenBank mRNA | AF226993.1 | - | 25 | 2777 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053792.1 | NP_446244.1 | 25 | 2777 | ||
GenBank mRNA | AF226993.1 | - | 25 | 2777 |