siRNA Details
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053801.2 | NP_446253.2 | 12 | 1198 | ||
XM_006251154.2 | XP_006251216.1 | 12 | 1187 | |||
GenBank mRNA | AF309558.1 | - | 12 | 1155 | ||
BC089785.1 | - | 12 | 1198 | 7322689 |
