siRNA Details
| Gene Symbol: Ebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053820.2 | NP_446272.2 | 15, 16 | 2014 | ||
| XM_006246117.2 | XP_006246179.1 | 15, 16 | 3619 | |||
| XM_006246118.2 | XP_006246180.1 | 15, 16 | 3616 | |||
| XM_006246120.2 | XP_006246182.1 | 16, 17 | 3373 | |||
| XM_008767631.1 | XP_008765853.1 | 15, 16 | 1970 | |||
| XM_008767632.1 | XP_008765854.1 | 15, 16 | 2384 | |||
| XM_008767633.1 | XP_008765855.1 | 13, 14 | 1505 | |||
| XM_008767634.1 | XP_008765856.1 | 12, 13 | 1867 | |||
| GenBank mRNA | FQ232013.1 | - | 2, 3 | 296 | ||
| L24051.1 | - | 15, 16 | 1756 | |||

Ebf1
Genomic Map