siRNA Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105746.1 | NP_001099216.1 | 4 | 1221 | ||
XM_006242298.2 | XP_006242360.1 | 10 | 1884 | |||
XM_006242301.2 | XP_006242363.1 | 9 | 2121 | |||
XM_008765654.1 | XP_008763876.1 | 9 | 2235 | |||
XM_008765655.1 | XP_008763877.1 | 10 | 1896 | |||
XM_008765656.1 | XP_008763878.1 | 10 | 1954 | |||
XM_008765657.1 | XP_008763879.1 | 9 | 2232 | |||
XM_008765658.1 | XP_008763880.1 | 9 | 2223 | |||
GenBank mRNA | L23204.1 | - | 5 | 1198 |
