siRNA Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053838.1 | NP_446290.1 | 13 | 2077 | ||
| XM_006238010.2 | XP_006238072.1 | 12 | 2717 | |||
| GenBank mRNA | M26896.1 | - | 13 | 2077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npr2| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053838.1 | NP_446290.1 | 13 | 2077 | ||
| XM_006238010.2 | XP_006238072.1 | 12 | 2717 | |||
| GenBank mRNA | M26896.1 | - | 13 | 2077 | ||