siRNA Details
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053840.2 | NP_446292.2 | 479 | |||
| GenBank mRNA | BC078768.1 | - | 479 | 7133071 | ||
| M33821.1 | - | 577 | ||||
| M33822.1 | - | 564 | ||||
| X15443.1 | - | 485 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggt1| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053840.2 | NP_446292.2 | 479 | |||
| GenBank mRNA | BC078768.1 | - | 479 | 7133071 | ||
| M33821.1 | - | 577 | ||||
| M33822.1 | - | 564 | ||||
| X15443.1 | - | 485 | ||||