siRNA Details
| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057108.2 | NP_476449.1 | 5, 6 | 683 | ||
| XM_008769405.1 | XP_008767627.1 | 4, 5 | 841 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb5| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057108.2 | NP_476449.1 | 5, 6 | 683 | ||
| XM_008769405.1 | XP_008767627.1 | 4, 5 | 841 | |||