siRNA Details
| Gene Symbol: Mapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053842.1 | NP_446294.1 | 7 | 1066 | ||
| XM_006248658.2 | XP_006248720.1 | 7 | 1103 | |||
| XM_006248659.2 | XP_006248721.1 | 7 | 1103 | |||
| XM_008768848.1 | XP_008767070.1 | 7 | 1103 | |||
| GenBank mRNA | FQ211642.1 | - | 7 | 1106 | ||
| M64300.1 | - | 7 | 1066 | |||

Mapk1
Genomic Map