siRNA Details
| Gene Symbol: Fcgr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053843.2 | NP_446295.2 | 5 | 687 | ||
| XM_006250303.2 | XP_006250365.1 | 5 | 740 | |||
| XM_006250305.2 | XP_006250367.1 | 5 | 839 | |||
| GenBank mRNA | BC060534.1 | - | 464 | 6886727 | ||
| L08446.1 | - | 5 | 687 | |||
| Gene Symbol: LOC103693683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769773.1 | XP_008767995.1 | 4 | 492 | ||

Fcgr2a
Genomic Map