siRNA Details
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053848.1 | NP_446300.1 | 1, 2 | 634 | ||
GenBank mRNA | M88709.1 | - | 784 | |||
M88710.1 | - | 1, 2 | 465 | |||
M88711.1 | - | 1, 2 | 634 |
