siRNA Details
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053848.1 | NP_446300.1 | 7 | 1415 | ||
XM_008766041.1 | XP_008764263.1 | 5 | 573 | |||
GenBank mRNA | M88709.1 | - | 1565 | |||
M88710.1 | - | 7 | 1246 | |||
M88711.1 | - | 7 | 1415 |
