siRNA Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058213.1 | NP_478120.1 | 12, 13 | 1587 | ||
| GenBank mRNA | BC168245.1 | - | 12, 13 | 1600 | 7935496 | |
| M99223.1 | - | 12, 13 | 1587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2a1| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058213.1 | NP_478120.1 | 12, 13 | 1587 | ||
| GenBank mRNA | BC168245.1 | - | 12, 13 | 1600 | 7935496 | |
| M99223.1 | - | 12, 13 | 1587 | |||