siRNA Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058213.1 | NP_478120.1 | 21 | 3084 | ||
| GenBank mRNA | BC168245.1 | - | 21 | 3097 | 7935496 | |
| FQ215287.1 | - | 6 | 805 | |||
| FQ215444.1 | - | 6 | 803 | |||
| FQ215994.1 | - | 6 | 805 | |||
| FQ215996.1 | - | 6 | 805 | |||
| FQ216214.1 | - | 6 | 658 | |||
| FQ216553.1 | - | 7 | 1053 | |||
| FQ216874.1 | - | 7 | 1152 | |||
| FQ217464.1 | - | 5 | 508 | |||
| M99223.1 | - | 21 | 3084 | |||

Atp2a1
Genomic Map