siRNA Details
| Gene Symbol: Nat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053854.1 | NP_446306.1 | 3 | 1146 | ||
| XM_006253007.1 | XP_006253069.1 | 2 | 2737 | |||
| XM_006253008.1 | XP_006253070.1 | 1 | 832 | |||
| XM_006253009.1 | XP_006253071.1 | 2 | 1924 | |||
| XM_008771063.1 | XP_008769285.1 | 2 | 911 | |||
| GenBank mRNA | U01348.1 | - | 3 | 1146 | ||
| U17261.1 | - | 1 | 841 | |||

Nat2
Genomic Map