siRNA Details
| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053859.2 | NP_446311.1 | 8 | 1110 | ||
| GenBank mRNA | EU253553.1 | - | 9 | 1075 | ||
| FQ211811.1 | - | 1123 | ||||
| U07609.1 | - | 8 | 1113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a7| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053859.2 | NP_446311.1 | 8 | 1110 | ||
| GenBank mRNA | EU253553.1 | - | 9 | 1075 | ||
| FQ211811.1 | - | 1123 | ||||
| U07609.1 | - | 8 | 1113 | |||