siRNA Details
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053864.2 | NP_446316.1 | 4 | 557 | ||
GenBank mRNA | BC060518.1 | - | 4 | 557 | 5597843 | |
U11760.1 | - | 4 | 589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053864.2 | NP_446316.1 | 4 | 557 | ||
GenBank mRNA | BC060518.1 | - | 4 | 557 | 5597843 | |
U11760.1 | - | 4 | 589 |