siRNA Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053866.3 | NP_446318.3 | 11 | 1649 | ||
| GenBank mRNA | CF110238.1 | - | 1 | 20 | ||
| CV110320.1 | - | 4 | 509 | |||
| FQ211509.1 | - | 1710 | ||||
| FQ223782.1 | - | 11 | 1724 | |||
| U17901.1 | - | 11 | 1508 | |||

Plaa
Genomic Map