siRNA Details
| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053868.2 | NP_446320.1 | 7 | 2886 | ||
| XM_008760890.1 | XP_008759112.1 | 4 | 1857 | |||
| GenBank mRNA | U22952.1 | - | 7 | 3250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlgn1| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053868.2 | NP_446320.1 | 7 | 2886 | ||
| XM_008760890.1 | XP_008759112.1 | 4 | 1857 | |||
| GenBank mRNA | U22952.1 | - | 7 | 3250 | ||