siRNA Details
| Gene Symbol: Srp54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053871.1 | NP_446323.1 | 14 | 1442 | ||
| GenBank mRNA | BC079117.1 | - | 14 | 1442 | 7126297 | |
| FQ211580.1 | - | 14 | 1453 | |||
| FQ225417.1 | - | 14 | 1457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp54a| Gene Symbol: Srp54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053871.1 | NP_446323.1 | 14 | 1442 | ||
| GenBank mRNA | BC079117.1 | - | 14 | 1442 | 7126297 | |
| FQ211580.1 | - | 14 | 1453 | |||
| FQ225417.1 | - | 14 | 1457 | |||