siRNA Details
| Gene Symbol: Srp54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053871.1 | NP_446323.1 | 9 | 944 | ||
| XM_006240097.2 | XP_006240159.1 | 9 | 956 | |||
| GenBank mRNA | BC079117.1 | - | 9 | 944 | 7126297 | |
| FQ211580.1 | - | 9 | 955 | |||
| FQ225417.1 | - | 9 | 959 | |||

Srp54a
Genomic Map