siRNA Details
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053879.1 | NP_446331.1 | 10 | 1283 | ||
XM_008763166.1 | XP_008761388.1 | 6 | 1014 | |||
GenBank mRNA | U35371.1 | - | 10 | 1283 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053879.1 | NP_446331.1 | 10 | 1283 | ||
XM_008763166.1 | XP_008761388.1 | 6 | 1014 | |||
GenBank mRNA | U35371.1 | - | 10 | 1283 |