siRNA Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053881.1 | NP_446333.1 | 18 | 2420 | ||
| XM_006245134.2 | XP_006245196.1 | 18 | 2491 | |||
| GenBank mRNA | D45414.1 | - | 10 | 1266 | ||
| FQ211652.1 | - | 18 | 2479 | |||
| U40652.1 | - | 18 | 2386 | |||
| X92563.1 | - | 18 | 2420 | |||

Ptprn
Genomic Map