siRNA Details
| Gene Symbol: Ecm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053882.1 | NP_446334.1 | 11 | 1640 | ||
| XM_006232942.2 | XP_006233004.1 | 11 | 1972 | |||
| GenBank mRNA | BC085879.1 | - | 11 | 1640 | 7191762 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ecm1| Gene Symbol: Ecm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053882.1 | NP_446334.1 | 11 | 1640 | ||
| XM_006232942.2 | XP_006233004.1 | 11 | 1972 | |||
| GenBank mRNA | BC085879.1 | - | 11 | 1640 | 7191762 | |