siRNA Details
| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053888.1 | NP_446340.1 | 16 | 3231 | ||
| GenBank mRNA | U48809.1 | - | 16 | 3251 | ||
| U67081.1 | - | 1339 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myt1l| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053888.1 | NP_446340.1 | 16 | 3231 | ||
| GenBank mRNA | U48809.1 | - | 16 | 3251 | ||
| U67081.1 | - | 1339 | ||||