siRNA Details
| Gene Symbol: Ppp1r12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053890.1 | NP_446342.1 | 24 | 2994 | ||
| XM_006241308.1 | XP_006241370.1 | 26 | 3437 | |||
| XM_006241309.1 | XP_006241371.1 | 27 | 3468 | |||
| XM_006241311.1 | XP_006241373.1 | 25 | 3323 | |||
| XM_006241312.1 | XP_006241374.1 | 25 | 3269 | |||
| XM_006241313.1 | XP_006241375.1 | 25 | 3257 | |||
| XM_006241314.1 | XP_006241376.1 | 26 | 3354 | |||
| XM_006241315.1 | XP_006241377.1 | 24 | 3143 | |||
| XM_006241316.2 | XP_006241378.1 | 24 | 3469 | |||
| GenBank mRNA | S74907.1 | - | 24 | 2994 | ||

Ppp1r12a
Genomic Map