siRNA Details
Gene Symbol: Ppp1r12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053890.1 | NP_446342.1 | 5 | 757 | ||
XM_006241308.1 | XP_006241370.1 | 5 | 918 | |||
XM_006241309.1 | XP_006241371.1 | 5 | 918 | |||
XM_006241311.1 | XP_006241373.1 | 5 | 918 | |||
XM_006241312.1 | XP_006241374.1 | 5 | 918 | |||
XM_006241313.1 | XP_006241375.1 | 5 | 918 | |||
XM_006241314.1 | XP_006241376.1 | 5 | 918 | |||
XM_006241315.1 | XP_006241377.1 | 5 | 918 | |||
XM_006241316.2 | XP_006241378.1 | 3 | 950 | |||
GenBank mRNA | FQ233443.1 | - | 5 | 923 | ||
S74907.1 | - | 5 | 757 | |||
U50185.1 | - | 5 | 637 |
