siRNA Details
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229825.2 | XP_006229887.1 | 6 | 1244 | ||
XM_006229830.2 | XP_006229892.1 | 6 | 1245 | |||
XM_006229831.2 | XP_006229893.1 | 6 | 1231 | |||
XM_006229832.2 | XP_006229894.1 | 6 | 1231 | |||
XM_006229834.2 | XP_006229896.1 | 5 | 1012 | |||
XM_006229835.2 | XP_006229897.1 | 3 | 1395 | |||
XM_006229837.2 | XP_006229899.1 | 5 | 1002 | |||
XM_006229838.2 | XP_006229900.1 | 5 | 1002 | |||
XM_008759662.1 | XP_008757884.1 | 3 | 795 | |||
XM_008759663.1 | XP_008757885.1 | 3 | 1336 | |||
GenBank mRNA | AB236144.1 | - | 4 | 477 | ||
U57715.1 | - | 5 | 994 |
