siRNA Details
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229825.2 | XP_006229887.1 | 8, 9 | 1781 | ||
XM_006229830.2 | XP_006229892.1 | 8, 9 | 1770 | |||
XM_006229831.2 | XP_006229893.1 | 8, 9 | 1593 | |||
XM_006229832.2 | XP_006229894.1 | 8, 9 | 1581 | |||
XM_006229834.2 | XP_006229896.1 | 7, 8 | 1549 | |||
XM_006229835.2 | XP_006229897.1 | 5, 6 | 1932 | |||
XM_006229837.2 | XP_006229899.1 | 7, 8 | 1364 | |||
XM_006229838.2 | XP_006229900.1 | 7, 8 | 1352 | |||
XM_008759662.1 | XP_008757884.1 | 5, 6 | 1332 | |||
XM_008759663.1 | XP_008757885.1 | 5, 6 | 1873 | |||
GenBank mRNA | AB236144.1 | - | 6, 7 | 839 | ||
FQ230423.1 | - | 1, 2 | 68 | |||
U57715.1 | - | 7, 8 | 1344 |
